Diethyl iminodiacetate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Iminodiacetic Acid Diethyl Ester, Diethyl 2,2'-azanediyldiacetate

H60608-AA H60608-AC
CAAAH60608-AAEA 53.88 CAD
CAAAH60608-AA CAAAH60608-AC
Diethyl iminodiacetate 97%
Diethyl iminodiacetate

Purity: 97% CAS No.: 6290-05-7 Molecular Formula: C8H15NO4 Formula Weight: 189.21 Flash Point: 113°(235°F) Refractive Index: 1.435 Boiling Point: 208° MDL No.: MFCD00041925

Formula: C₈H₁₅NO₄
MW: 189.21 g/mol
Boiling Pt: 208 °C
Density: 1.056
Flash Pt: 113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041925
CAS Number: 6290-05-7
EINECS: 228-533-6

Order Now

Specification Test Results

Assay Minimum 97

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR