4,6-O-Benzylidene-D-glucal 97%

Supplier: Thermo Scientific Chemicals

H60259-03
CAAAH60259-03EA 191.18 CAD
CAAAH60259-03 CAAAH60259-MD
4,6-O-Benzylidene-D-glucal 97%
4,6-O-Benzylidene-D-glucal

Purity: 97% CAS No.: 14125-70-3 Molecular Formula: C13H14O4 Formula Weight: 234.25 Storage Temperature: 2°C to 8°C MDL No.: MFCD00167506

Formula: C₁₃H₁₄O₄
Storage Temperature: Ambient
MDL Number: MFCD00167506
CAS Number: 14125-70-3

Order Now

Specification Test Results

Assay Minimum 97

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR