(R)-2,2'-Bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl ≥97%

Supplier: Thermo Scientific Chemicals
Warning

H60112-03 H60112-MD
CAAAH60112-03EA 1178.68 CAD
CAAAH60112-03 CAAAH60112-MD
(R)-2,2'-Bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl ≥97%
(R)-2,2'-Bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl

Purity: 97+% CAS No.: 1365531-98-1 Molecular Formula: C76H108O8P2 Formula Weight: 1211.61 MDL No.: MFCD20265636

Formula: C₇₆H₁₀₈O₈P₂
Storage Temperature: Ambient
MDL Number: MFCD20265636
CAS Number: 1365531-98-1

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR