1-Butyl-3-methylimidazolium dicyanamide 97%

Supplier: Thermo Scientific Chemicals
Warning

H59175-06 H59175-18
CAAAH59175-06EA 192.76 CAD
CAAAH59175-06 CAAAH59175-18
1-Butyl-3-methylimidazolium dicyanamide 97%
1-Butyl-3-methylimidazolium dicyanamide

CAS: 448245-52-1 MDL: MFCD07784457

Formula: C₁₀H₁₅N₅
Density: 1.06 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD07784457
CAS Number: 448245-52-1

Order Now

Specification Test Results

Assay Minimum 97

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR