N-(Trimethylsilyl)morpholine 97%

Supplier: Thermo Scientific Chemicals
Danger

H55229-06 H55229-14
CAAAH55229-03EA 62.84 CAD
CAAAH55229-03 CAAAH55229-06 CAAAH55229-14
N-(Trimethylsilyl)morpholine 97%
N-(Trimethylsilyl)morpholine

Purity: 97% CAS No.: 13368-42-8 Molecular Formula: C7H17NOSi Formula Weight: 159.30 Flash Point: 29°(84°F) Refractive Index: 1.4410 Boiling Point: 160° Beilstein Reference: 1071480 UN No.: 2924 MDL No.: MFCD00010098

Formula: C₇H₁₇NOSi
Boiling Pt: 160 °C
Density: 0.897
Flash Pt: 29 °C(84 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010098
CAS Number: 13368-42-8
UN: 2924
ADR: "3",III

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR