N,N'-Bis(2-aminoethyl)-1,3-propanediamine 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3,7-Diaza-1,9-nonanediamine, N1,N1'-(Propane-1,3-diyl)bis(ethane-1,2-diamine), 1,4,8,11-Tetraazaundecane

H54950-03 H54950-03
CAAAH54950-03EA 82.16 CAD
CAAAH54950-03 AAH54950-03
N,N'-Bis(2-aminoethyl)-1,3-propanediamine 96%
N,N'-Bis(2-aminoethyl)-1,3-propanediamine

Purity: 97% CAS No.: 4741-99-5 Molecular Formula: C7H20N4 Formula Weight: 160.26 Flash Point: 100°(212°F) Refractive Index: 1.494 Boiling Point: 142-145° Beilstein Reference: 1742732 UN No.: 2735 MDL No.: MFCD00008174

Formula: C₇H₂₀N₄
MW: 160.27 g/mol
Boiling Pt: 142…145 °C
Density: 0.96
Flash Pt: 100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008174
CAS Number: 4741-99-5
EINECS: 225-254-1
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR