Propargyl amine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Monopropagyl amine, Prop-2-ynylamine, 3-Aminopropyne, Prop-2-yn-1-amine

H53495-06
CAAAH53495-06EA 129.24 CAD
CAAAH53495-06
Propargyl amine 98%
Propagyl amine

Purity: 98% CAS No.: 2450-71-7 Molecular Formula: C3H5N Formula Weight: 55.08 Storage Temperature: 2°C to 8°C Flash Point: 6°(43°F) Refractive Index: 1.449 Boiling Point: 83° Beilstein Reference: 773681 UN No.: 3286 MDL No.: MFCD00008198

Formula: C₃H₅N
MW: 55.08 g/mol
Boiling Pt: 83 °C
Density: 0.86
Flash Pt: 6 °C (43 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008198
CAS Number: 2450-71-7
EINECS: 219-513-8
UN: 3286
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR