1H-Pyrrolo[2,3-b]pyridine-4-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H33139-03
CAAAH33139-03EA 330.22 CAD
CAAAH33139-03 CAAAH33139-MD
1H-Pyrrolo[2,3-b]pyridine-4-carboxylic acid 97%
1H-Pyrrolo[2,3-b]pyridine-4-carboxylic acid

Purity: 97% CAS No.: 479553-01-0 Molecular Formula: C8H6N2O2 Formula Weight: 162.15 Physical Form: Solid Appearance: White to cream Melting Point: 330° MDL No.: MFCD08272236

Formula: C₈H₆N₂O₂
Melting Pt: >330 °C
Storage Temperature: Ambient
MDL Number: MFCD08272236
CAS Number: 479553-01-0

Order Now

Specification Test Results

Assay Minimum 97

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR