4-Amino-2-methylbenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

H31657-03 H31657-06
CAAAH31657-03EA 25.75 CAD
CAAAH31657-03 CAAAH31657-06
4-Amino-2-methylbenzoic acid 98%
4-Amino-2-methylbenzoic acid

Purity: 98% CAS No.: 2486-75-1 Molecular Formula: C8H9NO2 Formula Weight: 151.16 Physical Form: Solid MDL No.: MFCD06656133

Formula: C₈H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD06656133
CAS Number: 2486-75-1

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR