(4-(Hydroxymethyl)phenyl)acetic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H27531-03 H27531-06
CAAAH27531-03EA 68.57 CAD
CAAAH27531-03 CAAAH27531-06
(4-(Hydroxymethyl)phenyl)acetic acid 97%
(4-(Hydroxymethyl)phenyl)acetic acid

Purity: 98% CAS No.: 73401-74-8 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 131-134° MDL No.: MFCD00065692

Formula: C₉H₁₀O₃
Melting Pt: 131…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00065692
CAS Number: 73401-74-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR