5-Bromo-1,2,3-trimethoxybenzene 97%

Supplier: Thermo Scientific Chemicals
Warning

B24872-06 B24872-14 B24872-22
CAAAB24872-06EA 67.31 CAD
CAAAB24872-06 CAAAB24872-14 CAAAB24872-22
5-Bromo-1,2,3-trimethoxybenzene 97%
5-Bromo-1,2,3-trimethoxybenzene

Purity: 97% CAS No.: 2675-79-8 Molecular Formula: C9H11BrO3 Formula Weight: 247.10 Melting Point: 77-80° MDL No.: MFCD00017169

Formula: C₉H₁₁BrO₃
Melting Pt: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00017169
CAS Number: 2675-79-8
EINECS: 220-223-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR