Isoeugenol cis- and trans-mixture 98+%

Supplier: Thermo Scientific Chemicals
Warning

B24541-14 B24541-22 B24541-36
CAAAB24541-14EA 38.39 CAD
CAAAB24541-14 CAAAB24541-22 CAAAB24541-36
Isoeugenol cis- and trans-mixture 98+%
Isoeugenol

Purity: 98+% CAS No.: 97-54-1 Molecular Formula: C10H12O2 Formula Weight: 164.21 Flash Point: 134°(273°F) Refractive Index: 1.5770 Melting Point: -10° Boiling Point: 266-268° Beilstein Reference: 1909602 Merck Reference: 14,5171 MDL No.: MFCD00009285

Formula: C₁₀H₁₂O₂
Boiling Pt: 266…268 °C
Melting Pt: –10 °C
Density: 1.080
Flash Pt: 134 °C(273 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009285
CAS Number: 97-54-1
EINECS: 202-590-7
Merck Index: 14,5171

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR