4-Chloro-3-sulfamoylbenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B24057-18 B24057-30
CAAAB24057-0BEA 814.62 CAD
CAAAB24057-0B CAAAB24057-18 CAAAB24057-30
4-Chloro-3-sulfamoylbenzoic acid 98%
4-Chloro-3-sulphamoylbenzoic acid

Purity: 98% CAS No.: 1205-30-7 Molecular Formula: C7H6ClNO4S Formula Weight: 235.65 Melting Point: 255-260° MDL No.: MFCD00012375

Formula: C₇H₆ClNO₄S
Melting Pt: 255…260 °C
Storage Temperature: Ambient
MDL Number: MFCD00012375
CAS Number: 1205-30-7
EINECS: 214-882-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR