Diethyl diethylmalonate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Diethyl Diethylmalonate

B23810-14 B23810-22 B23810-36
CAAAB23810-14EA 73.15 CAD
CAAAB23810-14 CAAAB23810-22 CAAAB23810-36
Diethyl diethylmalonate 98%
Diethyl diethylmalonate

Purity: 98% CAS No.: 77-25-8 Molecular Formula: C11H20O4 Formula Weight: 216.28 Flash Point: 94°(202°F) Refractive Index: 1.4230 Boiling Point: 228-230° Merck Reference: 14,3789 MDL No.: MFCD00009132

Formula: C₁₁H₂₀O₄
MW: 216.28 g/mol
Boiling Pt: 228…230 °C
Density: 0.990
Flash Pt: 94 °C(202 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009132
CAS Number: 77-25-8
EINECS: 201-016-2
Merck Index: 14,3789

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR