α-Bromo-2-chloro-4-fluorotoluene 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-(Bromomethyl)-2-chloro-4-fluorobenzene, 2-Chloro-4-fluorobenzyl bromide

B23565-06 B23565-14
CAAAB23565-06EA 29.39 CAD
CAAAB23565-06 CAAAB23565-14
α-Bromo-2-chloro-4-fluorotoluene 97%
α-Bromo-2-chloro-4-fluorotoluene

Purity: 97% CAS No.: 45767-66-6 Molecular Formula: C7H5BrClF Formula Weight: 223.48 Melting Point: 35-39° Beilstein Reference: 3539265 UN No.: 3261 MDL No.: MFCD00236025

Formula: C₇H₅BrClF
MW: 223.47
Melting Pt: 35…39 °C
Storage Temperature: Ambient
MDL Number: MFCD00236025
CAS Number: 45767-66-6
UN: 3261
ADR: 8 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR