Trimesic acid triethyl ester 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Triethyl 1,3,5-benzenetricarboxylate, Triethyl benzene-1,3,5-tricarboxylate

B23392-06 B23392-14
CAAAB23392-06EA 55.93 CAD
CAAAB23392-06 CAAAB23392-14 CAAAB23392-22
Trimesic acid triethyl ester 97%
Trimesic acid triethyl ester

Purity: 97% CAS No.: 4105-92-4 Molecular Formula: C15H18O6 Formula Weight: 294.31 Melting Point: 132-135° MDL No.: MFCD00236389

Formula: C₁₅H₁₈O₆
Melting Pt: 132…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00236389
CAS Number: 4105-92-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR