Daidzein 97%

Supplier: Thermo Scientific Chemicals
Warning

B22877-03 B22877-06 B22877-14
CAAAB22877-03EA 120.4 CAD
CAAAB22877-03 CAAAB22877-06 CAAAB22877-14
Daidzein 97%
Daidzein

Purity: 97% CAS No.: 486-66-8 Molecular Formula: C15H10O4 Formula Weight: 254.24 Melting Point: 328-332° dec. Beilstein Reference: 231523 Merck Reference: 14,2801 MDL No.: MFCD00016954

Formula: C₁₅H₁₀O₄
Boiling Pt: 513 °C (1013 hPa)
Melting Pt: 328…332 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00016954
CAS Number: 486-66-8
EINECS: 207-635-4
Merck Index: 14,2801

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR