Cystamine dihydrochloride 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cystaminium dichloride, 2,2′-Diaminodiethyl disulfide dihydrochloride, 2,2′-Diaminodiethyl disulphide dihydrochloride, 2,2'-Disulphanediyldiethanamine dihydrochloride, Bis(2-aminoethyl) disulphide dihydrochloride, {2-[(2-Aminoethyl)disulfanyl]ethyl}amine dihydrochloride, Decarboxycystine dihydrochloride, Cystamine (dihydrochloride), Bis(2-aminoethyl) disulfide dihydrochloride, {2-[(2-Aminoethyl)disulphanyl]ethyl}amine dihydrochloride, 2,2'-Disulfanediyldiethanamine dihydrochloride, 2,2′-Dithiobis(ethylamine) dihydrochloride

B22873-14 B22873-22
CAAAB22873-14EA 69.05 CAD
CAAAB22873-14 CAAAB22873-22
Cystamine dihydrochloride 97+%
Cystamine dihydrochloride

Cystamine dihydrochloride acts as an anti-infective agent, which is used in the treatment of urinary tract infections. It is also used as a radiation-protective agent that interferes with sulfhydryl enzymes. Further, it serves as a heparin antagonist and sulfhydryl modifying reagent. In addition to this, it is used as an inhibitor of TGase.


Purity: 97+% CAS No.: 56-17-7 Molecular Formula: C4H12N2S2.2HCl Formula Weight: 225.20 Odor: Odorless Melting Point: 217-220° Beilstein Reference: 3616850 Merck Reference: 14,2776 MDL No.: MFCD00012905

Formula: C₄H₁₄Cl₂N₂S₂
MW: 225.20 g/mol
Melting Pt: 217…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00012905
CAS Number: 56-17-7
EINECS: 200-260-7
Merck Index: 14,2776

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR