Sodium 1-Naphthalenesulfonate, (max. 2% H₂O) 99% (dry weight)

Supplier: Thermo Scientific Chemicals
Warning

B22488-06 B22488-14 B22488-22
CAAAB22488-06EA 59.57 CAD
CAAAB22488-06 CAAAB22488-14 CAAAB22488-22
Sodium 1-Naphthalenesulfonate, (max. 2% H₂O) 99% (dry weight)
Sodium 1-Naphthalenesulfonate

Purity: 99% CAS No.: 130-14-3 Molecular Formula: C10H7NaO3S Formula Weight: 230.22 Melting Point: 299-301° Beilstein Reference: 3639038 MDL No.: MFCD00064964

Formula: C₁₀H₈O₃S
Melting Pt: 299…301 °C
Storage Temperature: Ambient
MDL Number: MFCD00064964
CAS Number: 130-14-3
EINECS: 204-976-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR