Rhodanine-N-acetic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

B22244-06 B22244-14 B22244-22
CAAAB22244-06EA 47.24 CAD
CAAAB22244-06 CAAAB22244-14 CAAAB22244-22
Rhodanine-N-acetic acid 98%
Rhodanine-N-acetic acid

Purity: 98% CAS No.: 5718-83-2 Molecular Formula: C5H5NO3S2 Formula Weight: 191.23 Melting Point: 145-148° Beilstein Reference: 149513

Formula: C₅H₅NO₃S₂
Melting Pt: 145…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00005491
CAS Number: 5718-83-2
EINECS: 227-220-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR