2-Ethylacrolein 90% stabilized, tech.

Supplier: Thermo Scientific Chemicals
Danger


Stabilization: Stabilised with hydroquinone (HQ) 50 ppm

B21839-06 B21839-14 B21839-22
CAAAB21839-06EA 122.13 CAD
CAAAB21839-06 CAAAB21839-14 CAAAB21839-22
2-Ethylacrolein 90% stabilized, tech.
2-Ethylacrolein

Stab. with 50ppm hydroquinone


Purity: 90% CAS No.: 922-63-4 Molecular Formula: C5H8O Formula Weight: 84.12 Storage Temperature: 2°C to 8°C Flash Point: 1°(34°F) Refractive Index: 1.4245 Odor: Pungent Boiling Point: 92-93° Beilstein Reference: 1098378 UN No.: 1992 MDL No.: MFCD00010129

Formula: C₅H₈O
Boiling Pt: 92…93 °C
Density: 0.859
Flash Pt: 1 °C(34 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010129
CAS Number: 922-63-4
EINECS: 213-079-3
UN: 1992
ADR: "3",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR