Methyl-4-fluorobenzoate 97%

Supplier: Thermo Scientific Chemicals
Danger

B21504-06 B21504-14 B21504-22
CAAAB21504-06EA 90.85 CAD
CAAAB21504-06 CAAAB21504-14 CAAAB21504-22
Methyl-4-fluorobenzoate 97%
Methyl 4-fluorobenzoate

Purity: 97% CAS No.: 403-33-8 Molecular Formula: C8H7FO2 Formula Weight: 154.14 Flash Point: 77°(170°F) Refractive Index: 1.4950 Odor: Aromatic Boiling Point: 195-197° Beilstein Reference: 2085925 MDL No.: MFCD00017959

Formula: C₈H₇FO₂
Boiling Pt: 195…197 °C
Density: 1.192
Flash Pt: 77 °C(170 °F)
Storage Temperature: Ambient
MDL Number: MFCD00017959
CAS Number: 403-33-8
EINECS: 206-956-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR