Tetracycline hydrochloride 96%

Supplier: Thermo Scientific Chemicals
Warning

B21408-14 B21408-22
CAAAB21408-14EA 104.44 CAD
CAAAB21408-14 CAAAB21408-22
Tetracycline hydrochloride 96%
Tetracycline hydrochloride

Can induce apoptosis in osteoclasts


Purity: 96% CAS No.: 64-75-5 Molecular Formula: C22H24N2O8.HCl Formula Weight: 480.91 Melting Point: 220-223° Merck Reference: 14,9196 Optical Rotation: -247° (c=1, 0.1M HCl) MDL No.: MFCD00078142


Can induce apoptosis in osteoclasts

Formula: C₂₂H₂₅ClN₂O₈
Melting Pt: 220…223 °C
Storage Temperature: Ambient
MDL Number: MFCD00078142
CAS Number: 64-75-5
EINECS: 200-593-8
Merck Index: 14,9196

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR