Methyl-3-chlorobenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

B21371-14 B21371-22
CAAAB21371-14EA 70.78 CAD
CAAAB21371-14 CAAAB21371-22 CAAAB21371-36
Methyl-3-chlorobenzoate 98%
Methyl 3-chlorobenzoate

Purity: 98% CAS No.: 2905-65-9 Molecular Formula: C8H7ClO2 Formula Weight: 170.60 Flash Point: 104°(219°F) Refractive Index: 1.5310 Melting Point: 20-22° Boiling Point: 99-101°/12mm Beilstein Reference: 2045126 Merck Reference: 14,2123 MDL No.: MFCD00000592

Formula: C₈H₇ClO₂
Boiling Pt: 99…101 °C/12 mm
Melting Pt: 20…22 °C
Density: 1.227
Flash Pt: 104 °C(219 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000592
CAS Number: 2905-65-9
EINECS: 220-814-1
Merck Index: 14,2123

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR