6-Chloro-3-indolyl-β-D-galactopyranoside 98%

Supplier: Thermo Scientific Chemicals

B21131-02 B21131-77
CAAAB21131-02EA 519.82 CAD
CAAAB21131-02 CAAAB21131-77
6-Chloro-3-indolyl-β-D-galactopyranoside 98%
6-Chloro-3-indolyl-beta-D-galactopyranoside

Purity: 98% CAS No.: 138182-21-5 Molecular Formula: C14H16ClNO6 Formula Weight: 329.74 Storage Temperature: 2°C to 8°C Melting Point: 188-190° Optical Rotation: -47° (c=1 in methanol) MDL No.: MFCD00467206

Formula: C₁₄H₁₆ClNO₆
Melting Pt: 188…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00467206
CAS Number: 138182-21-5

Order Now

Specification Test Results

Optical rotation -47° (c=1 in methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR