Ethyl-7-methoxybenzofuran-2-carboxylat 97%

Supplier: Thermo Scientific Chemicals

B21066-03 B21066-06 B21066-14
CAAAB21066-03EA 37.6 CAD
CAAAB21066-03 CAAAB21066-06 CAAAB21066-14
Ethyl-7-methoxybenzofuran-2-carboxylat 97%
Ethyl 7-methoxybenzofuran-2-carboxylat

Purity: 97% CAS No.: 50551-58-1 Molecular Formula: C12H12O4 Formula Weight: 220.22 Melting Point: 83-86° MDL No.: MFCD00060510

Formula: C₁₂H₁₂O₄
Melting Pt: 83…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00060510
CAS Number: 50551-58-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR