Tetrakis(trimethylsilyl)silane 98%

Supplier: Thermo Scientific Chemicals
Warning

B21020-03 B21020-06
CAAAB21020-03EA 70.31 CAD
CAAAB21020-03 CAAAB21020-06
Tetrakis(trimethylsilyl)silane 98%
Tetrakis(trimethylsilyl)silane

Purity: 98% CAS No.: 4098-98-0 Molecular Formula: Si[Si(CH3)3]4 Formula Weight: 320.85 Melting Point: 266-268° subl. MDL No.: MFCD00054859

Formula: C₁₂H₃₆Si₅
Melting Pt: 266…268 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00054859
CAS Number: 4098-98-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR