(±)-3-Methyl-3-heptanol 98%

Supplier: Thermo Scientific Chemicals
Warning

B20959-06
CAAAB20959-03EA 54.59 CAD
CAAAB20959-03 CAAAB20959-06 CAAAB20959-14
(±)-3-Methyl-3-heptanol 98%
(±)-3-Methyl-3-heptanol

Purity: 98% CAS No.: 5582-82-1 Molecular Formula: C8H18O Formula Weight: 130.23 Flash Point: 58°(136°F) Boiling Point: 64°/201mm UN No.: 1987 MDL No.: MFCD00021841

Formula: C₈H₁₈O
Boiling Pt: 64 °C/201 mm
Density: 0.83
Flash Pt: 58 °C(136 °F)
Storage Temperature: Ambient
MDL Number: MFCD00021841
CAS Number: 5582-82-1
UN: 1987
ADR: "3",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR