α-Bromo-2,5-difluorotoluene 98%

Supplier: Thermo Scientific Chemicals
Danger

B20752-18 B20752-09
CAAAAB20752-18EA 554.58 CAD
CAAAAB20752-18 CAAAB20752-09
α-Bromo-2,5-difluorotoluene 98%
α-Bromo-2,5-difluorotoluene

Purity: 98% CAS No.: 85117-99-3 Molecular Formula: C7H5BrF2 Formula Weight: 207.02 Refractive Index: 1.5260 Beilstein Reference: 7089248 UN No.: 3265 MDL No.: MFCD00009897

Formula: C₇H₅BrF₂
Boiling Pt: 186 °C (1013 hPa)
Melting Pt: 8 °C
Density: 1.609
Storage Temperature: Ambient
MDL Number: MFCD00009897
CAS Number: 85117-99-3
EINECS: 285-651-0
UN: 2924
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR