5-(4-Pyridyl)-1H-tetrazole 98%

Supplier: Thermo Scientific Chemicals

B20741-03
CAAAB20741-03EA 108.23 CAD
CAAAB20741-03 CAAAB20741-06
5-(4-Pyridyl)-1H-tetrazole 98%
5-(4-Pyridyl)-1H-tetrazole

Purity: 98% CAS No.: 14389-12-9 Molecular Formula: C6H5N5 Formula Weight: 147.14 MDL No.: MFCD00068115

Formula: C₆H₅N₅
Storage Temperature: Ambient
MDL Number: MFCD00068115
CAS Number: 14389-12-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR