(-)-Carveol (mixture of cis- and trans-isomers) 97%

Supplier: Thermo Scientific Chemicals

B20636-09 B20636-18 B20636-30
CAAAB20636-09EA 85.48 CAD
CAAAB20636-09 CAAAB20636-18 CAAAB20636-30
(-)-Carveol (mixture of cis- and trans-isomers) 97%
(-)-Carveol (mixture of cis- and trans-isomers)

Purity: 97% CAS No.: 99-48-9 Molecular Formula: C10H16O Formula Weight: 152.24 Flash Point: 98°(209°F) Refractive Index: 1.4960 Boiling Point: 227° Beilstein Reference: 6269416 MDL No.: MFCD00869995

Formula: C₁₀H₁₆O
Boiling Pt: 227 °C
Density: 0.96
Flash Pt: 98 °C(209 °F)
Storage Temperature: Ambient
MDL Number: MFCD00869995
CAS Number: 99-48-9
EINECS: 202-757-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR