N,N'-Bis(2-hydroxyethyl)oxamide 99%

Supplier: Thermo Scientific Chemicals

B20458-06 B20458-14 B20458-22
CAAAB20458-06EA 68.57 CAD
CAAAB20458-06 CAAAB20458-14 CAAAB20458-22
N,N'-Bis(2-hydroxyethyl)oxamide 99%
N,N'-Bis(2-hydroxyethyl)oxamide

Purity: 99% CAS No.: 1871-89-2 Molecular Formula: C6H12N2O4 Formula Weight: 176.17 Melting Point: 167-169° dec. MDL No.: MFCD00020559

Formula: C₆H₁₂N₂O₄
Melting Pt: 167…169 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00020559
CAS Number: 1871-89-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR