4-Acetamidobenzaldehyde 98%

Supplier: Thermo Scientific Chemicals

B20430-06 B20430-14 B20430-22
CAAAB20430-06EA 67.31 CAD
CAAAB20430-06 CAAAB20430-14 CAAAB20430-22
4-Acetamidobenzaldehyde 98%
4-Acetamidobenzaldehyde

Purity: 98% CAS No.: 122-85-0 Molecular Formula: C9H9NO2 Formula Weight: 163.18 Melting Point: 154-157° MDL No.: MFCD00003380

Formula: C₉H₉NO₂
Melting Pt: 154…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00003380
CAS Number: 122-85-0
EINECS: 204-579-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR