3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B20412-06 B20412-14
CAAAB20412-06EA 191.18 CAD
CAAAB20412-06 CAAAB20412-14
3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid 98%
3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid

Purity: 98% CAS No.: 20170-32-5 Molecular Formula: C17H26O3 Formula Weight: 278.40 Melting Point: 172-175° MDL No.: MFCD00017519

Formula: C₁₇H₂₆O₃
Melting Pt: 172…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00017519
CAS Number: 20170-32-5
EINECS: 243-556-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR