2-Methyl-5-hexen-3-ol 99%

Supplier: Thermo Scientific Chemicals
Warning

B20220-06 B20220-14
CAAAB20220-03EA 147.54 CAD
CAAAB20220-03 CAAAB20220-06 CAAAB20220-14
2-Methyl-5-hexen-3-ol 99%
2-Methyl-5-hexen-3-ol

Purity: 99% CAS No.: 32815-70-6 Molecular Formula: C7H14O Formula Weight: 114.19 UN No.: 1987 MDL No.: MFCD00048326

Formula: C₇H₁₄O
MW: 114.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00048326
CAS Number: 32815-70-6
UN: 1987
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR