(3-Bromophenyl)acetonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

B20189-22 B20189-06 B20189-14
CAAAAB20189-22EA 725.22 CAD
CAAAAB20189-22 CAAAB20189-06 CAAAB20189-14
(3-Bromophenyl)acetonitrile 97%
(3-Bromophenyl)acetonitrile

Purity: 97% CAS No.: 31938-07-5 Molecular Formula: C8H6BrN Formula Weight: 196.05 Flash Point: 126°(259°F) Refractive Index: 1.5700 Melting Point: 25-28° Boiling Point: 275° Beilstein Reference: 2355439 UN No.: 3449 MDL No.: MFCD00001906

Formula: C₈H₆BrN
Boiling Pt: 275 °C
Melting Pt: 25…28 °C
Flash Pt: 126 °C (259 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001906
CAS Number: 31938-07-5
EINECS: 250-867-6
UN: 3449
ADR: 6.1,I

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR