1H,1H,2H,2H-Tridecafluoro-1-octanol 97%

Supplier: Thermo Scientific Chemicals
Warning

B20156-30 B20156-09 B20156-18
CAAAAB20156-30EA 2362.1 CAD
CAAAAB20156-30 CAAAB20156-09 CAAAB20156-18
1H,1H,2H,2H-Tridecafluoro-1-octanol 97%
1H,1H,2H,2H-Tridecafluoro-1-octanol

Purity: 97% CAS No.: 647-42-7 Molecular Formula: CF3(CF2)5(CH2)2OH Formula Weight: 364.11 Flash Point: 91°(195°F) Refractive Index: 1.3140 Boiling Point: 90°/20mm Beilstein Reference: 1992757 MDL No.: MFCD00042143

Formula: C₈H₅F₁₃O
Boiling Pt: 90 °C/20 mm
Density: 1.68
Flash Pt: 91 °C(195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042143
CAS Number: 647-42-7
EINECS: 211-477-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR