2,4,6-Trimethyl-1,6-heptadien-4-ol 98%

Supplier: Thermo Scientific Chemicals

B20086-03 B20086-06 B20086-14
CAAAB20086-03EA 45.82 CAD
CAAAB20086-03 CAAAB20086-06 CAAAB20086-14
2,4,6-Trimethyl-1,6-heptadien-4-ol 98%
2,4,6-Trimethyl-1,6-heptadien-4-ol

Purity: 98% CAS No.: 79604-66-3 Molecular Formula: C10H18O Formula Weight: 154.25 MDL No.: MFCD00026060

Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00026060
CAS Number: 79604-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR