(Methoxycarbonylsulfamoyl)triethylammonium hydroxide inner salt 96%

Supplier: Thermo Scientific Chemicals
Warning

L20155-03 L20155-06 L20155-14
CAAAAL20155-03EA 129.09 CAD
CAAAAL20155-03 CAAAAL20155-06 CAAAAL20155-14
(Methoxycarbonylsulfamoyl)triethylammonium hydroxide inner salt 96%
(Methoxycarbonylsulfamoyl)triethylammonium hydroxide inner salt

Fieser: 14,208 21,266


Purity: 96% CAS No.: 29684-56-8 Molecular Formula: C8H18N2O4S Formula Weight: 238.30 Storage Temperature: -30°C to -10°C Melting Point: 72-82° Beilstein Reference: 1432131 MDL No.: MFCD00077815

Formula: C₈H₁₈N₂O₄S
Melting Pt: 72…82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077815
CAS Number: 29684-56-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR