Ethyl-2-methyl-3-hydroxy-4,4,4-trifluorobutyrate 97%

Supplier: Thermo Scientific Chemicals
Warning

L19969-06
CAAAAL19969-03EA 120 CAD
CAAAAL19969-03 CAAAAL19969-06
Ethyl-2-methyl-3-hydroxy-4,4,4-trifluorobutyrate 97%
Ethyl-2-methyl-3-hydroxy-4,4,4-trifluorobutyrate

Purity: 97% CAS No.: 91600-33-8 Molecular Formula: C7H11F3O3 Formula Weight: 200.16 Boiling Point: 180-184° Beilstein Reference: 6194166 MDL No.: MFCD00190634

Formula: C₇H₁₁F₃O₃
MW: 200.16 g/mol
Boiling Pt: 180…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00190634
CAS Number: 91600-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR