4-Amino-3-nitropyridine 96%

Supplier: Thermo Scientific Chemicals
Warning

L19725-06 L19725-14
CAAAAL19725-03EA 51.3 CAD
CAAAAL19725-03 CAAAAL19725-06 CAAAAL19725-14
4-Amino-3-nitropyridine 96%
4-Amino-3-nitropyridine

Purity: 96% CAS No.: 1681-37-4 Molecular Formula: C5H5N3O2 Formula Weight: 139.11 Melting Point: 201-205° Beilstein Reference: 383710

Formula: C₅H₅N₃O₂
Melting Pt: 201…205 °C
Storage Temperature: Ambient
MDL Number: MFCD00160304
CAS Number: 1681-37-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR