1H-Pyrazole-4-boronic acid pinacol ester 98%

Supplier: Thermo Scientific Chemicals
Warning

L19654-03 L19654-06
CAAAAL19654-03EA 120.24 CAD
CAAAAL19654-03 CAAAAL19654-06
1H-Pyrazole-4-boronic acid pinacol ester 98%
1H-Pyrazole-4-boronic acid pinacol ester

Purity: 98% CAS No.: 269410-08-4 Molecular Formula: C9H15BN2O2 Formula Weight: 194.04 Melting Point: 145-149° MDL No.: MFCD03453063

Formula: C₉H₁₅BN₂O₂
Melting Pt: 145…149 °C
Storage Temperature: Ambient
MDL Number: MFCD03453063
CAS Number: 269410-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR