tert-Butyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate 97%

Supplier: Thermo Scientific Chemicals
Warning

L19653-03 L19653-06
CAAAAL19653-03EA 96.54 CAD
CAAAAL19653-03 CAAAAL19653-06
tert-Butyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate 97%
tert-Butyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate

Purity: 97% CAS No.: 330793-01-6 Molecular Formula: C17H26BNO4 Formula Weight: 319.21 Melting Point: 169-170° MDL No.: MFCD02179439

Formula: C₁₇H₂₆BNO₄
Melting Pt: 169…170 °C
Storage Temperature: Ambient
MDL Number: MFCD02179439
CAS Number: 330793-01-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR