2,3-Dibromopyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

L19527-06 L19527-14
CAAAAL19527-03EA 60.96 CAD
CAAAAL19527-03 CAAAAL19527-06 CAAAAL19527-14
2,3-Dibromopyridine 97%
2,3-Dibromopyridine

Purity: 97% CAS No.: 13534-89-9 Molecular Formula: C5H3Br2N Formula Weight: 236.90 Melting Point: 58-60° Boiling Point: 249-250° MDL No.: MFCD00234014

Formula: C₅H₃Br₂N
MW: 236.89 g/mol
Boiling Pt: 249…250 °C
Melting Pt: 58…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00234014
CAS Number: 13534-89-9
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR