3,5-Difluorobenzenesulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

L19507-06
CAAAAL19507-03EA 96.82 CAD
CAAAAL19507-03 CAAAAL19507-06
3,5-Difluorobenzenesulfonyl chloride 97%
3,5-Difluorobenzenesulphonyl chloride

Purity: 97% CAS No.: 210532-25-5 Molecular Formula: C6H3ClF2O2S Formula Weight: 212.60 Flash Point: 110°(230°F) Melting Point: 57-59° Beilstein Reference: 8053561 UN No.: 3261 MDL No.: MFCD02091378

Formula: C₆H₃ClF₂O₂S
Melting Pt: 57…59 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD02091378
CAS Number: 210532-25-5
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR