(S)-(+)-4-Methyl-2-pentanol 99%

Supplier: Thermo Scientific Chemicals
Warning

L18882-03 L18882-MD
CAAAAL18882-03EA 319.16 CAD
CAAAAL18882-03 CAAAAL18882-MD
(S)-(+)-4-Methyl-2-pentanol 99%
(S)-(+)-4-Methyl-2-pentanol

For synthesis of optically active products


Purity: 99% CAS No.: 14898-80-7 Molecular Formula: C6H14O Formula Weight: 102.18 Flash Point: 41°(105°F) Refractive Index: 1.4110 Odor: Slightly sweetish Boiling Point: 130-131° Beilstein Reference: 1718991 Optical Rotation: +21° (neat) UN No.: 2053 MDL No.: MFCD03093078

Formula: C₆H₁₄O
Boiling Pt: 130…131 °C
Density: 0.802
Flash Pt: 41 °C (105 °F)
Storage Temperature: Ambient
MDL Number: MFCD03093078
CAS Number: 14898-80-7
UN: 2053
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR