1,2,4-Triazolo[4,3-a]pyridin-3(2H)-one ≥98%

Supplier: Thermo Scientific Chemicals
Warning

L18705-06 L18705-14
CAAAAL18705-06EA 41.4 CAD
CAAAAL18705-06 CAAAAL18705-14
1,2,4-Triazolo[4,3-a]pyridin-3(2H)-one ≥98%
1,2,4-Triazolo[4,3-a]pyridin-3(2H)-one

Purity: 98+% CAS No.: 6969-71-7 Molecular Formula: C6H5N3O Formula Weight: 135.13 Melting Point: 233-235° Beilstein Reference: 607433 MDL No.: MFCD00022632

Formula: C₆H₅N₃O
Melting Pt: 233…235 °C
Storage Temperature: Ambient
MDL Number: MFCD00022632
CAS Number: 6969-71-7
EINECS: 230-191-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR