3,5-Bis(trifluoromethyl)-o-phenylenediamine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3,5-Bis(trifluoromethyl)-1,2-diaminobenzene

L18180-03 L18180-06
CAAAAL18180-03EA 89.59 CAD
CAAAAL18180-03 CAAAAL18180-06
3,5-Bis(trifluoromethyl)-o-phenylenediamine 97%
3,5-Bis(trifluoromethyl)-o-phenylenediamine

Purity: 97% CAS No.: 367-65-7 Molecular Formula: C8H6F6N2 Formula Weight: 244.14 Melting Point: 44-49° Beilstein Reference: 3344332 UN No.: 2811 MDL No.: MFCD01631430

Formula: C₈H₆F₆N₂
MW: 244.14 g/mol
Melting Pt: 44…49 °C
Storage Temperature: Ambient
MDL Number: MFCD01631430
CAS Number: 367-65-7
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR