4-(Methylsulfinylphenyl)boronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

L17865-03 L17865-MC
CAAAAL17865-03EA 360.24 CAD
CAAAAL17865-03 CAAAAL17865-MC
4-(Methylsulfinylphenyl)boronic acid 98%
4-(Methylsulfinylphenyl)boronic acid

Purity: 98% CAS No.: 166386-48-7 Molecular Formula: C7H9BO3S Formula Weight: 184.02 Melting Point: ca 170° MDL No.: MFCD02093071

Formula: C₇H₉BO₃S
Melting Pt: ca 170 °C
Storage Temperature: Ambient
MDL Number: MFCD02093071
CAS Number: 166386-48-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR