6-Bromoindole 98%

Supplier: Thermo Scientific Chemicals
Warning

L17703-03 L17703-MD
CAAAAL17703-03EA 301.78 CAD
CAAAAL17703-03 CAAAAL17703-MD
6-Bromoindole 98%
6-Bromoindole

Purity: 98% CAS No.: 52415-29-9 Molecular Formula: C8H6BrN Formula Weight: 196.05 Melting Point: 91-93° Boiling Point: 70-75°/0.01mm MDL No.: MFCD00238550

Formula: C₈H₆BrN
Boiling Pt: 70…75 °C/0.01 mm
Melting Pt: 91…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00238550
CAS Number: 52415-29-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR